COMH4211B021 Combi-Blocks, Inc.

1-(Cyclopropylmethyl)-1h-indole-4-carboxylic acid hydrazide

  • MDL number MFCD31539564
  • Cas Number : 1035927-98-0
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3355

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3355
Availability details for
Quantity
  • Availability
  • Add to Cart