COMH4211B022 Combi-Blocks, Inc.

Butanoic acid, 4-(3-acetylphenoxy)-, ethyl ester

  • MDL number MFCD31381658
  • Cas Number : 756485-01-5
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3357

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3357
Availability details for
Quantity
  • Availability
  • Add to Cart