COMH4211B023 Combi-Blocks, Inc.

5-Chloro-2-(2-oxo-oxazolidin-3-yl)-benzonitrile

  • MDL number MFCD28402081
  • Cas Number : 1192348-50-7
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3359
Availability details for
Quantity
  • Availability
  • Add to Cart