COMH4211B027 Combi-Blocks, Inc.

2-[[[(1,1-Dimethylethyl)dimethylsilyl]oxy]methyl]-2-methylpropanal

  • MDL number MFCD09868649
  • Cas Number : 144681-67-4
  • Molecular Weight : 216.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3365

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3365
Availability details for
Quantity
  • Availability
  • Add to Cart