COMH4211B028 Combi-Blocks, Inc.

2-Oxetan-3-yl-propionic acid ethyl ester

  • MDL number MFCD31618959
  • Cas Number : 1467674-35-6
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3366

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3366
Availability details for
Quantity
  • Availability
  • Add to Cart