COMH4211B02B Combi-Blocks, Inc.

2-Bromo-3-isobutoxy-benzaldehyde

  • MDL number MFCD31613853
  • Cas Number : 1999788-51-0
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3374
Availability details for
Quantity
  • Availability
  • Add to Cart