COMH4211B02C Combi-Blocks, Inc.

(4-Bromo-3-chlorophenyl)(ethyl)sulfane

  • MDL number MFCD22483683
  • Cas Number : 1005206-32-5
  • Molecular Weight : 251.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3377
Availability details for
Quantity
  • Availability
  • Add to Cart