COMH4211B02E Combi-Blocks, Inc.

4-(Tetrahydro-pyran-4-ylmethyl)-phenylamine

  • MDL number MFCD31614067
  • Cas Number : 1780609-31-5
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3382
Availability details for
Quantity
  • Availability
  • Add to Cart