COMH4211B02F Combi-Blocks, Inc.

4-Propoxy-1-naphthaldehyde

  • MDL number MFCD06248739
  • Cas Number : 54784-09-7
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3383

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3383
Availability details for
Quantity
  • Availability
  • Add to Cart