COMH4211B030 Combi-Blocks, Inc.

4-(4-Carboxy-benzyl)-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD31614022
  • Cas Number : 2270913-32-9
  • Molecular Weight : 353.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3386
Availability details for
Quantity
  • Availability
  • Add to Cart