COMH4211B031 Combi-Blocks, Inc.

(3-Cyclopropylmethoxy-5-hydroxymethyl-phenyl)-methanol

  • MDL number MFCD31628813
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3387
Availability details for
Quantity
  • Availability
  • Add to Cart