COMH4211B032 Combi-Blocks, Inc.

1-(2,4-Difluoro-benzyl)-3,5-dimethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD31583216
  • Molecular Weight : 348.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3390

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3390
Availability details for
Quantity
  • Availability
  • Add to Cart