COMH4211B033 Combi-Blocks, Inc.

(R)-2-((2,2-Dimethyl-1,3-dioxolan-4-yl)methoxy)nicotinic acid

  • MDL number MFCD31628823
  • Cas Number : 1219446-76-0
  • Molecular Weight : 253.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-2-((2,2-dimethyl-1,3-dioxolan-4-yl)methoxy)nicotinic acid
Inchi Code 1S/C12H15NO5/c1-12(2)17-7-8(18-12)6-16-10-9(11(14)15)4-3-5-13-10/h3-5,8H,6-7H2,1-2H3,(H,14,15)/t8-/m1/s1
Inchi Key FKRCGPLCZBPVIA-MRVPVSSYSA-N

Description

Purity 95%
Country of Origin CN
Availability details for
Quantity
  • Availability
  • Add to Cart