COMH4211B034 Combi-Blocks, Inc.

Trans 4-(2-methoxycarbonyl-vinyl)-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD31557799
  • Cas Number : 239073-59-7
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3392

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3392
Availability details for
Quantity
  • Availability
  • Add to Cart