COMH4211B035 Combi-Blocks, Inc.

4-(4-Aminophenoxy)picolinic acid

  • MDL number MFCD23378603
  • Cas Number : 1012058-77-3
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3393

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3393
Availability details for
Quantity
  • Availability
  • Add to Cart