COMH4211B036 Combi-Blocks, Inc.

3-Methoxy-2,2-dimethylpropionic acid chloromethyl ester

  • MDL number MFCD31583274
  • Cas Number : 1350552-00-9
  • Molecular Weight : 180.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3397
Availability details for
Quantity
  • Availability
  • Add to Cart