COMH4211B037 Combi-Blocks, Inc.

tert-Butyl[3-(2-amino-5-chlorophenoxy)benzyl]carbamate

  • MDL number MFCD31579477
  • Cas Number : 266368-58-5
  • Molecular Weight : 348.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3398
Availability details for
Quantity
  • Availability
  • Add to Cart