COMH4211B038 Combi-Blocks, Inc.

(5-Chloropyrazin-2-yl)(pyrrolidin-1-yl)methanone

  • MDL number MFCD28099495
  • Cas Number : 1245215-68-2
  • Molecular Weight : 211.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-chloropyrazin-2-yl)(pyrrolidin-1-yl)methanone
Inchi Code 1S/C9H10ClN3O/c10-8-6-11-7(5-12-8)9(14)13-3-1-2-4-13/h5-6H,1-4H2
Inchi Key ZJSNMXJHOCFSNJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3399
Availability details for
Quantity
  • Availability
  • Add to Cart