COMH4211B039 Combi-Blocks, Inc.

1-(Cyclopropylmethyl)pyrrolidin-3-ol

  • MDL number MFCD17248940
  • Cas Number : 331281-31-3
  • Molecular Weight : 141.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3400
Availability details for
Quantity
  • Availability
  • Add to Cart