COMH4211B03A Combi-Blocks, Inc.

Pentanoic acid, 5-[[(2r)-2-[[(1,1-dimethylethoxy)carbonyl]amino]-1-oxo-3-[(triphenylmethyl)thio]propyl]amino]-

  • MDL number MFCD31539513
  • Cas Number : 1100364-96-2
  • Molecular Weight : 562.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3405

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3405
Availability details for
Quantity
  • Availability
  • Add to Cart