COMH4211B03B Combi-Blocks, Inc.

3-Chloro-4-(ethanesulfonyl)aniline

  • MDL number MFCD09938463
  • Cas Number : 1016887-45-8
  • Molecular Weight : 219.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3409
Availability details for
Quantity
  • Availability
  • Add to Cart