COMH4211B03C Combi-Blocks, Inc.

(3-Amino-4-fluorophenyl)(pyrrolidin-1-yl)methanone

  • MDL number MFCD11936259
  • Cas Number : 1155912-44-9
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3410
Availability details for
Quantity
  • Availability
  • Add to Cart