COMH4211B03E Combi-Blocks, Inc.

N-[(2,4-Dichlorophenyl)methyl]cyclopropanamine

  • MDL number MFCD07408436
  • Cas Number : 892568-95-5
  • Molecular Weight : 216.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2,4-dichlorobenzyl)cyclopropanamine
Inchi Code 1S/C10H11Cl2N/c11-8-2-1-7(10(12)5-8)6-13-9-3-4-9/h1-2,5,9,13H,3-4,6H2
Inchi Key ISNLISSGALOLGC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3412
Availability details for
Quantity
  • Availability
  • Add to Cart