COMH4211B03F Combi-Blocks, Inc.

tert-Butyl 3-((3-bromobenzyloxy)carbonylamino)propanoate

  • MDL number MFCD31583355
  • Cas Number : 1404431-59-9
  • Molecular Weight : 358.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3415
Availability details for
Quantity
  • Availability
  • Add to Cart