COMH4211B043 Combi-Blocks, Inc.

3-[(5-Fluoro-2-methylbenzyl)oxy]azetidine

  • MDL number MFCD11875490
  • Cas Number : 1121624-25-6
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3428
Availability details for
Quantity
  • Availability
  • Add to Cart