COMH4211B044 Combi-Blocks, Inc.

4-(Benzyloxy)cyclohexyl methanesulfonate

  • MDL number MFCD26391241
  • Cas Number : 100864-60-6
  • Molecular Weight : 284.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3429

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3429
Availability details for
Quantity
  • Availability
  • Add to Cart