COMH4211B045 Combi-Blocks, Inc.

1,1-Dimethyl-2,3-dihydro-1h-indene

  • MDL number MFCD11616489
  • Cas Number : 4912-92-9
  • Molecular Weight : 146.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3430
Availability details for
Quantity
  • Availability
  • Add to Cart