COMH4211B046 Combi-Blocks, Inc.

N-(4-Bromo-3-fluoro-benzoyl)-methanesulfonamide

  • MDL number MFCD31561466
  • Cas Number : 1971914-64-3
  • Molecular Weight : 296.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3431
Availability details for
Quantity
  • Availability
  • Add to Cart