COMH4211B047 Combi-Blocks, Inc.

1-(4-Bromobutoxy)-2-fluorobenzene

  • MDL number MFCD02030537
  • Cas Number : 106558-68-3
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3433
Availability details for
Quantity
  • Availability
  • Add to Cart