COMH4211B049 Combi-Blocks, Inc.

4-Iodo-1-(4-nitrophenyl)-1h-pyrazole

  • MDL number MFCD12031020
  • Cas Number : 153142-44-0
  • Molecular Weight : 315.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3436
Availability details for
Quantity
  • Availability
  • Add to Cart