COMH4211B04B Combi-Blocks, Inc.

1-(3-Chloro-4-cyclopropylmethoxy-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31539532
  • Cas Number : 2205505-15-1
  • Molecular Weight : 262.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3442
Availability details for
Quantity
  • Availability
  • Add to Cart