COMH4211B04D Combi-Blocks, Inc.

1-Bromo-4-chloro-2-(difluoromethoxy)benzene

  • MDL number MFCD14698391
  • Cas Number : 1056942-37-0
  • Molecular Weight : 257.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3448

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3448
Availability details for
Quantity
  • Availability
  • Add to Cart