COMH4211B04F Combi-Blocks, Inc.

2-(4-Chloro-2-fluorophenoxy)-2-methylpropanoic acid

  • MDL number MFCD27934599
  • Cas Number : 952483-86-2
  • Molecular Weight : 232.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3450

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3450
Availability details for
Quantity
  • Availability
  • Add to Cart