COMH4211B050 Combi-Blocks, Inc.

2-(5-Phenyl-oxazol-2-yl)-phenylamine

  • MDL number MFCD00489663
  • Cas Number : 193811-53-9
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3454
Availability details for
Quantity
  • Availability
  • Add to Cart