COMH4211B051 Combi-Blocks, Inc.

2-Methyl-3-propoxy-propan-1-ol

  • MDL number MFCD31618961
  • Cas Number : 2139805-63-1
  • Molecular Weight : 132.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-3-propoxypropan-1-ol

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3456

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3456
Availability details for
Quantity
  • Availability
  • Add to Cart