COMH4211B052 Combi-Blocks, Inc.

4'-Bromo-4-methanesulfonyl-biphenyl

  • MDL number MFCD13176584
  • Cas Number : 1774-28-3
  • Molecular Weight : 311.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3458
Availability details for
Quantity
  • Availability
  • Add to Cart