COMH4211B053 Combi-Blocks, Inc.

2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazol-1-yl]cyclopentanol

  • MDL number MFCD22573076
  • Cas Number : 1313726-16-7
  • Molecular Weight : 278.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3465
Availability details for
Quantity
  • Availability
  • Add to Cart