COMH4211B054 Combi-Blocks, Inc.

5-Bromo-2-(4-methoxy-benzyloxy)-phenylamine

  • MDL number MFCD11147551
  • Cas Number : 1020944-10-8
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3466
Availability details for
Quantity
  • Availability
  • Add to Cart