COMH4211B056 Combi-Blocks, Inc.

2-Chloro-2'-fluorobiphenyl-4-amine

  • MDL number MFCD31583298
  • Cas Number : 1628016-02-3
  • Molecular Weight : 221.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3469

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3469
Availability details for
Quantity
  • Availability
  • Add to Cart