COMH4211B057 Combi-Blocks, Inc.

Sulindac sulfone

  • MDL number MFCD00871143
  • Cas Number : 59864-04-9
  • Molecular Weight : 372.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3471
Availability details for
Quantity
  • Availability
  • Add to Cart