COMH4211B058 Combi-Blocks, Inc.

3-(2-Fluoro-4-methyl-phenyl)-5-methyl-isoxazole-4-carboxylic acid ethyl ester

  • MDL number MFCD22370416
  • Cas Number : 1159602-14-8
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3474
Availability details for
Quantity
  • Availability
  • Add to Cart