COMH4211B05B Combi-Blocks, Inc.

3-(4,4-Dimethyl-piperidin-1-yl)-propionic acid hydrochloride

  • MDL number MFCD31579335
  • Molecular Weight : 221.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3481
Availability details for
Quantity
  • Availability
  • Add to Cart