COMH4211B05C Combi-Blocks, Inc.

2-Amino-4-(2-methoxy-ethoxy)-benzoic acid

  • MDL number MFCD24500050
  • Cas Number : 860530-65-0
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3482
Availability details for
Quantity
  • Availability
  • Add to Cart