COMH4211B05D Combi-Blocks, Inc.

[5-Bromo-2-(2-methoxy-ethoxy)-phenyl]-pyrrolidin-1-yl-methanone

  • MDL number MFCD07627434
  • Cas Number : 904875-65-6
  • Molecular Weight : 328.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[5-bromo-2-(2-methoxyethoxy)benzoyl]pyrrolidine
Inchi Code 1S/C14H18BrNO3/c1-18-8-9-19-13-5-4-11(15)10-12(13)14(17)16-6-2-3-7-16/h4-5,10H,2-3,6-9H2,1H3
Inchi Key XCGGCWKOAWVLSW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3485
Availability details for
Quantity
  • Availability
  • Add to Cart