COMH4211B060 Combi-Blocks, Inc.

1-(Methoxymethyl)-1h-indazole-5-carboxylic acid

  • MDL number MFCD28663093
  • Cas Number : 1404449-16-6
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3490
Availability details for
Quantity
  • Availability
  • Add to Cart