COMH4211B061 Combi-Blocks, Inc.

N-(Cyclopropylmethyl)-4-fluoroaniline

  • MDL number MFCD03211302
  • Cas Number : 356539-43-0
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3493
Availability details for
Quantity
  • Availability
  • Add to Cart