COMH4211B063 Combi-Blocks, Inc.

4-Bromo-2'-ethoxybiphenyl

  • MDL number MFCD14582787
  • Cas Number : 1443352-08-6
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3496
Availability details for
Quantity
  • Availability
  • Add to Cart