COMH4211B064 Combi-Blocks, Inc.

4-Acetyl-2-trifluoromethyl-benzoic acid

  • MDL number MFCD31613818
  • Cas Number : 2270905-74-1
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3499
Availability details for
Quantity
  • Availability
  • Add to Cart