COMH4211B067 Combi-Blocks, Inc.

3-(3-Fluoro-4-methylphenyl)-2-methylpropanoic acid

  • MDL number MFCD23756130
  • Cas Number : 1522334-06-0
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3506

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3506
Availability details for
Quantity
  • Availability
  • Add to Cart