COMH4211B06D Combi-Blocks, Inc.

Methyl 4-hydroxy-5-nitro-2-(trifluoromethyl)benzoate

  • MDL number MFCD31561602
  • Cas Number : 1163281-01-3
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3526

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3526
Availability details for
Quantity
  • Availability
  • Add to Cart